Learn More
6,7-Dimethoxyquinazoline-2,4-dione, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 28888-44-0 |
| Molecular Formula | C10H10N2O4 |
| MDL Number | MFCD00023889 |
| Quantity | 5 g |
| SMILES | O=C(N1)NC2=C(C=C(OC)C(OC)=C2)C1=O |
| Molecular Weight (g/mol) | 222.20 |
| Percent Purity | 97% |
| Chemical Name or Material | 6,7-Dimethoxyquinazoline-2,4-dione |