missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-methyluracil, 97%, Thermo Scientific™

Product Code. 11427136
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427136 25 g 25g
11437136 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427136 Supplier Thermo Scientific Alfa Aesar Supplier No. B25448.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.

Solubility
Soluble in diluted sodium hydroxide solution.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2434-53-9
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00075366
InChI Key GZLZRPNUDBIQBM-UHFFFAOYSA-N
Synonym 6-amino-1-methyluracil, 6-amino-1-methylpyrimidine-2,4 1h,3h-dione, 1-methyl-6-aminouracil, 2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl, 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-amino-1-methyl-2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl-1h-pyrimidine-2,4-dione, 6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione, 6-amino-1-methyl-3h-pyrimidine-2,4-dione, zlchem 437
PubChem CID 75520
IUPAC Name 6-amino-1-methylpyrimidine-2,4-dione
SMILES CN1C(=O)NC(=O)C=C1N

Specifications

Melting Point >300°C
Beilstein 127771
Solubility Information Soluble in diluted sodium hydroxide solution.
Quantity 25 g
Odor Odorless
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 6-Amino-1-methyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.