missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline, 98%

Product Code. 11373267
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373267 1 g 1g
11383267 5 g 5g
11393267 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373267 Supplier Thermo Scientific Alfa Aesar Supplier No. L13819.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 580-15-4 | C9H8N2 | 144.177 g/mol

6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-15-4
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006803
InChI Key RJSRSRITMWVIQT-UHFFFAOYSA-N
Synonym 6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
PubChem CID 11373
IUPAC Name quinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2)N)N=C1

Specifications

Melting Point 115°C to 119°C
Boiling Point 146°C (0.3 mmHg)
Odor Odorless
Quantity 1 g
Beilstein 113320
Sensitivity Air Sensitive
Solubility Information Soluble in methanol,chloroform,ethanol,and benzene. Insoluble in water.
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 6-Aminoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.