Learn More
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 356560-80-0 |
| Molecular Formula | C6H4BrN3 |
| MDL Number | MFCD08689534 |
| Quantity | 1 g |
| SMILES | BrC1=CN2C(C=C1)=NC=N2 |
| Molecular Weight (g/mol) | 198.02 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Bromo-[1,2,4]triazolo[1,5-a]pyridine |