missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoic acid, 98+%

Product Code. 11462418
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11462418

Brand: Thermo Scientific Alfa Aesar A15761.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4224-70-8 | C6H11BrO2 | 195.056 g/mol

6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

Solubility
Soluble in methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4224-70-8
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004422
InChI Key NVRVNSHHLPQGCU-UHFFFAOYSA-N
Synonym 6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
PubChem CID 20210
ChEBI CHEBI:60700
IUPAC Name 6-bromohexanoic acid
SMILES C(CCC(=O)O)CCBr

Specifications

Melting Point 32°C to 35°C
Boiling Point 165°C to 170°C (20 mmHg)
Flash Point 95°C (203°F)
Odor Odorless
Quantity 100 g
UN Number UN3261
Beilstein 1749740
Solubility Information Soluble in methanol.
Formula Weight 195.06
Percent Purity ≥98%
Chemical Name or Material 6-Bromohexanoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.