missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-methyl-4-phenyl-2(1H)-quinazolinone, TRC

Product Code. 30133685
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133685 500 mg 500mg
17923555 50 mg 50mg
17951445 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133685 Supplier Toronto Research Chemicals Supplier No. TRCC370030500MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20927-53-1
Molecular Formula C15 H11 Cl N2 O
Molecular Weight (g/mol) 270.71
Synonym 2(1H)-Quinazolinone, 6-chloro-1-methyl-4-phenyl-, 6-Chloro-1-methyl-4-phenyl-2(1H)-quinazolinone, 1-Methyl-4-phenyl-6-chloro-2(1H)-quinazolinone, 1-Methyl-4-phenyl-6-chloro-2-quinazolinone, 6-Chloro-1-methyl-4-phenyl-2(1H)-quinazolone, 6-Chloro-4-phenyl-1-methyl-2(1H)-quinazolinone
IUPAC Name 6-chloro-1-methyl-4-phenylquinazolin-2-one
SMILES CN1C(=O)N=C(c2ccccc2)c3cc(Cl)ccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H11ClN2O/c1-18-13-8-7-11(16)9-12(13)14(17-15(18)19)10-5-3-2-4-6-10/h2-9H,1H3
Formula Weight 270.056
Percent Purity >95%
Chemical Name or Material 6-Chloro-1-methyl-4-phenyl-2(1H)-quinazolinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.