missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,4-dimethoxypyrimidine, 98+%

Product Code. 11450287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450287 25 g 25g
11430287 1 g 1g
11440287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21696.14
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6320-15-6 | C6H7ClN2O2 | 174.58 g/mol

It is used in the synthesis of silyl and stannyl pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of silyl and stannyl pyrimidines.

Solubility
Soluble in chloroform, ethyl acetate and methanol.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-15-6
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00006068
InChI Key JHNRTJRDRWKAIW-UHFFFAOYSA-N
Synonym 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine
PubChem CID 80600
IUPAC Name 4-chloro-2,6-dimethoxypyrimidine
SMILES COC1=CC(Cl)=NC(OC)=N1

Specifications

Melting Point 71°C to 73°C
Quantity 25 g
Beilstein 137581
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform,ethyl acetate and methanol.
Formula Weight 174.59
Percent Purity ≥98%
Chemical Name or Material 6-Chloro-2,4-dimethoxypyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.