missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxaldehyde, TRC

Product Code. 30136651
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136651 50 mg 50mg
30133967 10 mg 10mg
17911355 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136651 Supplier Toronto Research Chemicals Supplier No. TRCC36520050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93955-15-8
Molecular Formula C15 H8 Cl2 N2 O
Molecular Weight (g/mol) 303.14
Synonym 6-Chloro-4-(2-chlorophenyl)-quinazoline-2-carbaldehyde, Lorazepam Imp. E (EP)
IUPAC Name 6-chloro-4-(2-chlorophenyl)quinazoline-2-carbaldehyde
SMILES Clc1ccc2nc(C=O)nc(c3ccccc3Cl)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H8Cl2N2O/c16-9-5-6-13-11(7-9)15(19-14(8-20)18-13)10-3-1-2-4-12(10)17/h1-8H
Formula Weight 302.0014
Chemical Name or Material 6-Chloro-4-(2-chlorophenyl)-quinazoline-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.