missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorobenzo[d]oxazole-2(3H)-thione, 98%, BLD Pharm™

Product Code. 30942998
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30942998 1 g 1g
30940687 100 g 100g
30939350 25 g 25g
30941855 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30942998 Supplier BLD Pharm Supplier No. BD1087011g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 22876-20-6
Molecular Formula C7H4ClNOS
MDL Number MFCD00800495
Quantity 1 g
SMILES S=C1OC2=CC(Cl)=CC=C2N1
Molecular Weight (g/mol) 185.63
Percent Purity 98%
Chemical Name or Material 6-Chlorobenzo[d]oxazole-2(3H)-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.