Learn More
6-chloroimidazo[2,1-b][1,3]thiazole, 97%, Thermo Scientific™
Product Code. 10713833
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10713833 1 g 1g
10311645 5 g 5g
10019591 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10713833 Supplier Thermo Scientific Maybridge Supplier No. SPB05749DA
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 23576-81-0
Molecular Formula C5H3ClN2S
Molecular Weight (g/mol) 158.603
MDL Number MFCD00173802
InChI Key WOZMDYAJHVHPMD-UHFFFAOYSA-N
PubChem CID 721582
IUPAC Name 6-chloroimidazo[2,1-b][1,3]thiazole
SMILES C1=CSC2=NC(=CN21)Cl
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5CH2CH2NCS
Quantity 1 g
Synonym 6-chloroimidazo 2,1-b thiazole, 6-chloroimidazo 2,1-b 1,3 thiazole, 6-chloroimidazole 2,1-b 1,3 thiazole, 6-chloroimidazo 2,1-b 1,3-thiazoline, chloroimidazobthiazole, acmc-1cip3, 6-chloro-imidazo 2,1-b thiazole, imidazo 2,1-b thiazole,6-chloro, imidazo 2,1-b thiazole, 6-chloro, 6-chloro-4h-imidazo 2,1-b thiazole
Formula Weight 158.61
Percent Purity 97%
Chemical Name or Material 6-chloroimidazo[2, 1-b][1, 3]thiazole