missing translation for 'onlineSavingsMsg'
Learn More

6-(chloromethyl)benzothiazole, 97%, Thermo Scientific™

Product Code. 11956791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11956791 5 g 5g
11946791 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956791 Supplier Thermo Scientific Acros Supplier No. 441350050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 149440-49-3
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.66
InChI Key WVJSICZOZHJCLJ-UHFFFAOYSA-N
Synonym 6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
PubChem CID 22905771
IUPAC Name 6-(chloromethyl)-1,3-benzothiazole
SMILES C1=CC2=C(C=C1CCl)SC=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 183.66
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-(Chloromethyl)benzothiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-(Chloromethyl)benzothiazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.