missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-carboxylic Acid, TRC

Product Code. 30353013
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30353013 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353013 Supplier Toronto Research Chemicals Supplier No. TRCC3806155G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Chloro-2-picolinic acid
CAS 4684-94-0
Molecular Formula C6 H4 Cl N O2
Quantity 5 g
Synonym Picolinic acid, 6-chloro- (6CI,7CI,8CI), 6-Chloro-2-pyridinecarboxylic acid, 2-Chloropyridine-6-carboxylic acid, 6-Chloro-3-picolinic acid, 6-Chloropicolinic acid, NSC 51587
InChI Formula InChI=1S/C6H4ClNO2/c7-5-3-1-2-4(8-5)6(9)10/h1-3H,(H,9,10)
SMILES OC(=O)c1cccc(Cl)n1
IUPAC Name 6-chloropyridine-2-carboxylic acid
Molecular Weight (g/mol) 157.55
Formula Weight 156.9931
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.