missing translation for 'onlineSavingsMsg'
Learn More

6-Cyanonicotinic acid, 97%

Product Code. 11333966
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333966 250 mg 250mg
11323966 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333966 Supplier Thermo Scientific Alfa Aesar Supplier No. L17127.MD
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 70165-31-0 | C7H4N2O2 | 148.121 g/mol

6-Cyanonicotinic acid, is used in medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Cyanonicotinic acid, is used in medicine

Solubility
Soluble in alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70165-31-0
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD01318135
InChI Key WMHSQCDPPJRWIL-UHFFFAOYSA-N
Synonym 6-cyanonicotinic acid, 2-cyano-5-carboxypyridine, 6-cyanonicotinicacid, 3-pyridinecarboxylic acid, 6-cyano, 5-carboxypicolinonitrile, 3-pyridinecarboxylicacid, 6-cyano, 6-cyano-3-pyridinecarboxylic acid, 6-cyano-nicotinic acid, pubchem16844, 6-cyanonicotinic acid,
PubChem CID 2761108
IUPAC Name 6-cyanopyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)C#N

Specifications

Melting Point 189°C to 194°C
Quantity 250 mg
Beilstein 4384027
Solubility Information Soluble in alcohol.
Formula Weight 148.12
Percent Purity 97%
Chemical Name or Material 6-Cyanonicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.