missing translation for 'onlineSavingsMsg'
Learn More

6-Dimethylaminopurine, TRC

Product Code. 30012736
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012736 250 mg 250mg
30005990 500 mg 500mg
30011201 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012736 Supplier Toronto Research Chemicals Supplier No. TRCD461900250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-55-6
Molecular Formula C7 H9 N5
Molecular Weight (g/mol) 163.18
Synonym 9H-Purin-6-amine, N,N-dimethyl- (ACI), 1H-Purin-6-amine, N,N-dimethyl- (9CI), Adenine, N,N-dimethyl- (8CI), Adenine, N6,N6-dimethyl- (6CI), N,N-Dimethyl-9H-purin-6-amine (ACI), Purine, 6-(dimethylamino)- (7CI), 6,6-Dimethyladenine, 6-(Dimethylamino)purine, 6-DMAP, Dimethyladenine, DMAP, N,N-Dimethyl-6-aminopurine, N,N-Dimethyladenine, N6,N6-Dimethyladenine, NSC 401568
IUPAC Name N,N-dimethyl-7H-purin-6-amine
SMILES CN(C)c1ncnc2nc[nH]c12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H9N5/c1-12(2)7-5-6(9-3-8-5)10-4-11-7/h3-4H,1-2H3,(H,8,9,10,11)
Formula Weight 163.0858
Chemical Name or Material 6-Dimethylaminopurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.