Learn More
6-Fluoro-8-Quinolinesulfonyl Chloride, TRC
Product Code. 30391198
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391198 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30391198 Supplier Toronto Research Chemicals Supplier No. TRCF59655025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Fluoro-8-Quinolinesulfonyl Chloride
CAS 1016515-89-1
Molecular Formula C9H5ClFNO2S
Quantity 25 mg
InChI Formula InChI=1S/C9H5ClFNO2S/c10-15(13,14)8-5-7(11)4-6-2-1-3-12-9(6)8/h1-5H
SMILES O=S(=O)(Cl)C1=CC(F)=CC2=CC=CN=C21
IUPAC Name 6-fluoroquinoline-8-sulfonyl chloride
Molecular Weight (g/mol) 245.66
Formula Weight 244.97