Learn More
6-Hydroxy-1,4-benzodioxane, TRC
Tekniske data
Specifications
| Chemical Name or Material | 6-Hydroxy-1,4-benzodioxane |
| CAS | 10288-72-9 |
| Molecular Formula | C8H8O3 |
| Quantity | 100 mg |
| Synonym | 2,3-Dihydro-1,4-benzodioxin-6-ol, 1,4-Benzodioxan-6-ol, 2,3-Dihydro-6-hydroxy-1,4-benzodioxin, 2,3-Dihydrobenzo[b][1,4]dioxin-6-ol |
| InChI Formula | InChI=1S/C8H8O3/c9-6-1-2-7-8(5-6)11-4-3-10-7/h1-2,5,9H,3-4H2 |
| SMILES | Oc1ccc2OCCOc2c1 |
| IUPAC Name | 2,3-dihydro-1,4-benzodioxin-6-ol |
| Molecular Weight (g/mol) | 152.15 |
| Formula Weight | 152.0473 |