missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-methoxy-7-deazapurine, TRC

Product Code. 30389603
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389603 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389603 Supplier Toronto Research Chemicals Supplier No. TRCH946325250MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 6-Hydroxy-2-methoxy-7-deazapurine
CAS 90057-07-1
Molecular Formula C7 H7 N3 O2
Quantity 250 mg
InChI Formula InChI=1S/C7H7N3O2/c1-12-7-9-5-4(2-3-8-5)6(11)10-7/h2-3H,1H3,(H2,8,9,10,11)
SMILES COc1nc2nccc2c(O)[nH]1
IUPAC Name 2-methoxy-3H-pyrrolo[2,3-d]pyrimidin-4-ol
Molecular Weight (g/mol) 165.15
Formula Weight 165.0538
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.