missing translation for 'onlineSavingsMsg'
Learn More

6’-Hydroxy Buspirone, TRC

Product Code. 18021840
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18021840 5 mg 5mg
18032920 10 mg 10mg
18070410 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18021840 Supplier Toronto Research Chemicals Supplier No. TRCH8308005MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125481-61-0
Molecular Formula C21 H31 N5 O3
Molecular Weight (g/mol) 401.5
Synonym 8-Azaspiro[4.5]decane-7,9-dione, 6-hydroxy-8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, 6-Hydroxy-8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione, 6-Hydroxybuspirone, 6'-Hydroxybuspirone, BMS 528215, BMY 28674
IUPAC Name 10-hydroxy-8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione
SMILES OC1C(=O)N(CCCCN2CCN(CC2)c3ncccn3)C(=O)CC14CCCC4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H31N5O3/c27-17-16-21(6-1-2-7-21)18(28)19(29)26(17)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20/h5,8-9,18,28H,1-4,6-7,10-16H2
Formula Weight 401.2427
Percent Purity >95%
Chemical Name or Material 6-Hydroxy Buspirone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.