Learn More
6-Hydroxybenzo[d][1,3]oxathiol-2-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4991-65-5 |
| Molecular Formula | C7H4O3S |
| MDL Number | MFCD00005859 |
| Quantity | 1 g |
| SMILES | OC1=CC=C2SC(=O)OC2=C1 |
| Molecular Weight (g/mol) | 168.17 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Hydroxybenzo[d][1,3]oxathiol-2-one |