Learn More
6-Methoxy-1-tetralone, TRC
Product Code. 30472444
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472444 Supplier Toronto Research Chemicals Supplier No. TRCM26869025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Methoxy-1-tetralone
CAS 1078-19-9
Molecular Formula C11H12O2
Quantity 25 g
Synonym 3,4-Dihydro-6-methoxy-1(2H)-naphthalenone, 1,2,3,4-Tetrahydro-6-methoxy-1-naphthalenone, 1-Oxo-1,2,3,4-tetrahydro-6-methoxynaphthalene, 6-Methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-Methoxy-1-oxo-1,2,3,4-tetrahydronaphthalene, 6-Methoxy-3,4-dihydro-1(2H)-naphthalenone, 6-Methoxy-3,4-dihydro-2H-naphthalen-1-one, 6-Methoxy-3,4-dihydronaphthalene-1(2H)-one, 6-Methoxy-α-tetralone, NSC 41700
InChI Formula InChI=1S/C11H12O2/c1-13-9-5-6-10-8(7-9)3-2-4-11(10)12/h5-7H,2-4H2,1H3
SMILES COc1ccc2C(=O)CCCc2c1
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837