missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxynicotinonitrile, 97%, Thermo Scientific™

Product Code. 10397204
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10397204 25 g 25g
10477442 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10397204 Supplier Thermo Scientific Maybridge Supplier No. SPB04785EB
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 15871-85-9
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00084981
InChI Key DFPYAQAFVHRSAG-UHFFFAOYSA-N
Synonym 6-methoxynicotinonitrile, 2-methoxypyridine-5-carbonitrile, 6-methoxy-3-pyridinecarbonitrile, 2-methoxy-5-cyanopyridine, 5-cyano-2-methoxypyridine, 2-methoxy-5-pyridinecarbonitrile, 6-methoxy-nicotinonitrile, 3-pyridinecarbonitrile, 6-methoxy, 6-metoxynicotinonitrile, pubchem15154
PubChem CID 2736776
IUPAC Name 6-methoxypyridine-3-carbonitrile
SMILES COC1=NC=C(C=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 6-Methoxynicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.