missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-thiouracil, TRC

Product Code. 30464534
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464534 25 g 25g
30464822 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464534 Supplier Toronto Research Chemicals Supplier No. TRCM33135525G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-04-2
Molecular Formula C5 H6 N2 O S
Molecular Weight (g/mol) 142.18
Synonym 4(1H)-Pyrimidinone, 2,3-dihydro-6-methyl-2-thioxo-, Uracil, 6-methyl-2-thio- (8CI), 2-Mercapto-4-methyl-6-hydroxypyrimidine, 2-Mercapto-6-methyl-4-pyrimidinol, 2-Mercapto-6-methylpyrimidin-4-one, 2-Mercapto-6-methylpyrimidyl-4-one, 2-Thio-6-methyluracil, 4-Hydroxy-2-mercapto-6-methylpyrimidine, 4-Hydroxy-6-methyl-2-mercaptopyrimidine, 4-Hydroxy-6-methylpyrimidine-2(1H)-thione, 4-Methyl-2-thiouracil, 6-Methyl-2-mercaptouracil, 6-Methyl-2-thio-2,4-(1H,3H)-pyrimidinedione, 6-Methyl-2-thiouracil, 6-Methyl-4-oxo-2-thioxo-1,2,3,4-tetrahydropyrimidine, 6-Thio-4-methyluracil, Alkiron, Antibason, Basecil, Basethyrin, MTU, Metacil, Methacil, Methiacil, Methicil, Methiocil, Methylthiouracil, Metiur, Muracil, Muracin, NSC 193526, NSC 314271, NSC 9378, Orcanon, Prostrumyl, Strumacil, Thimecil, Thiomecil, Thiomidil, Thioryl, Thiothymin, Thiothyron, Thiuryl, Thyreonorm, Thyril, Tiomeracil, Tiorale M, Tiotiron, USAF-EK 6454
IUPAC Name 6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CC1=CC(=O)NC(=S)N1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H6N2OS/c1-3-2-4(8)7-5(9)6-3/h2H,1H3,(H2,6,7,8,9)
Formula Weight 142.0201
Chemical Name or Material 6-Methyl-2-thiouracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.