missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,6-diazabicyclo[3.1.1]heptane Dihydrochloride, TRC

Product Code. 30464452
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464452 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464452 Supplier Toronto Research Chemicals Supplier No. TRCM302845500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-3,6-diazabicyclo[3.1.1]heptane Dihydrochloride
CAS 2231676-00-7
Molecular Formula C6 H12 N2 . 2[H Cl]
Quantity 500 mg
InChI Formula InChI=1S/C6H12N2.ClH/c1-8-5-2-6(8)4-7-3-5;/h5-7H,2-4H2,1H3;1H
SMILES Cl.CN1C2CNCC1C2
IUPAC Name 6-methyl-3,6-diazabicyclo[3.1.1]heptane;hydrochloride
Molecular Weight (g/mol) 148.63382
Formula Weight 148.0767
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.