Learn More
6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Product Code. p-7077478 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15264348 250 mg 250mg
15254348 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15264348 Supplier Thermo Scientific Acros Supplier No. 454392500
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610768-32-6
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD08669608
InChI Key JEECGGXGHJUYMN-UHFFFAOYSA-N
Synonym 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-picoline-5-boronic acid pinacol ester, 6-methylpyridine-3-boronic acid pinacol ester, 2-picoline-5-boronic acid pinacolate, 2-methylpyridine-5-boronic acid, pinacol ester, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-picoline-5-boronic acidpinacolate, 2-methylpyridine-5-boronic acid pinacolate
PubChem CID 16414272
IUPAC Name 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Specifications
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 219.09
Percent Purity 95%
Chemical Name or Material 6-Methylpyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 6-Methylpyridine-3-boronic acid pinacol ester, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.