Learn More
6-Methylquinazoline-2,4(1H,3H)-dione, 97%, Thermo Scientific™
Product Code. 15499858
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499858 5 g 5g
15409868 250 mg 250mg
15489858 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15499858 Supplier Thermo Scientific Alfa Aesar Supplier No. H63259.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62484-16-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD00831482
InChI Key OXVPNGBFUAIXTE-UHFFFAOYSA-N
Synonym 6-methylquinazoline-2,4 1h,3h-dione, 6-methylquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-methyl, 6-methyl-1h-quinazoline-2,4-quinone, 6-methyl-2,4 1h,3h-quinazolinedione, 6-methylquinazoline-2,4-1h,3h-dione, 2,4-dihydroxy-6-methylquinazoline, quinazoline-2,4-dione,6-me, 6-methyl-2,4-1h,3h-quinazolinedione, 2,4 1h,3h-quinazolinedione,6-methyl
PubChem CID 334024
IUPAC Name 6-methyl-1H-quinazoline-2,4-dione
SMILES CC1=CC=C2NC(=O)NC(=O)C2=C1
Specifications
Color White
Quantity 5 g
Formula Weight 176.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Methylquinazoline-2,4(1H,3H)-dione