missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-2-thiouracil, TRC

Product Code. 17983435
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983435 1 g 1g
17961535 50 mg 50mg
17972875 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983435 Supplier Toronto Research Chemicals Supplier No. TRCP3371051G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36822-11-4
Molecular Formula C10 H8 N2 O S
Molecular Weight (g/mol) 204.248
Synonym Uracil, 6-phenyl-2-thio- (6CI,7CI,8CI), 2,3-Dihydro-6-phenyl-2-thioxo-4(1H)-pyrimidinone, 2-Mercapto-6-phenylpyrimidin-4-ol, 6-Phenyl-2-thiouracil, 6-Phenylthiouracil, Fenucil, NSC 163991, NSC 42600
IUPAC Name 6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES O=C1NC(=S)NC(=C1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H8N2OS/c13-9-6-8(11-10(14)12-9)7-4-2-1-3-5-7/h1-6H,(H2,11,12,13,14)
Formula Weight 204.0357
Chemical Name or Material 6-Phenyl-2-thiouracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.