Learn More
(6-Phenyl-3-pyridinyl)methanol, 97%, Thermo Scientific™
Product Code. 10021604
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10021604 250 mg 250mg
10755915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10021604 Supplier Thermo Scientific Maybridge Supplier No. CC21109CB
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Chemical Identifiers
CAS 4634-09-7
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.23
MDL Number MFCD03086132
InChI Key RXXIVABOKBNIEI-UHFFFAOYSA-N
Synonym 6-phenylpyridin-3-yl methanol, 6-phenyl-3-pyridinyl methanol, 3-pyridinemethanol, 6-phenyl, 6-phenyl-3-pyridylmethanol, 3-pyridinemethanol,6-phenyl, 6-phenyl-3-pyridyl-methanol, 2-phenyl-5-hydroxymethylpyridine, rxxivabokbniei-uhfffaoysa-n, 6-phenyl-3-pyridyl methan-1-ol
PubChem CID 22254216
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 185.23
Percent Purity 97%
Chemical Name or Material (6-Phenyl-3-pyridinyl)methanol