Learn More
Labconco 6 place Microplate Rotor - CentriVap Pro
Product Code. 30533408 Shop All Labconco Corporation Products
Change view
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Material (Coating)
30533408 Each -
30533433 Each PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30533408 Supplier Labconco Supplier No. 7555900
  • ONLINE EXCLUSIVE PRICE
    2235.00€
    2072.65€ / Pack of 1
    Save 162.35€
    7% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stainless steel material
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Rotor System CentriVap Pro
Dimensions (L x W x H) 28.45 x 27.69 x 8.13 cm (11.2 x 10.9 x 3.2 in.)