Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Frontier™ R-A12x5/14MS Rotors
Product Code. 15866632 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
6 x 1.0 mL
6 x 1.8 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15866632 6 x 1.0 mL Pack of 6
15876632 6 x 1.8 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15866632 Supplier Ohaus™ Supplier No. 30130887
Only null left
Add to basket
Add to basket
Facilitate centrifugation of tubes smaller than the original, provided carrier space for Frontier™ R-A12x5/14MS Rotors
  • Allows you to centrifuge tubes that are smaller in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-A12x5/14MS Rotors
TRUSTED_SUSTAINABILITY
Specifications
Capacity 6 x 1.0 mL
Max. Tube Diameter 12.6 mm
For Use With (Equipment) R-A12x5/14MS Rotor
Description Adapter, D12.6mm, 1.0ml Cryo x6
No. of Places 1
No. per Pack 6