Description
- Allows you to centrifuge tubes that are smaller in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-A12x5/14MS Rotors
Specifications
Specifications
| Capacity | 6 x 1.8 mL |
| Max. Tube Diameter | 12.6 mm |
| For Use With (Equipment) | R-A12x5/14MS Rotor |
| Description | Adapter, D12.6mm, 1.8ml Cryo x6 |
| No. of Places | 1 |
| No. per Pack | 6 |