Learn More
3M™ 6000 Series Full Face Mask Respirators
Description
- Large lens for wide field of view and excellent visibility
- Well-balanced design with silicone face seal for greater comfort and durability
- Unique center adapter directs exhaled breath and moisture downward, reducing valve debris and facilitating quick and easy cleaning
Specifications
Specifications
| Certifications/Compliance | EN 136:1998 (Class 1) |
| Connection Type | Bayonet |
| Color | Light Gray |
| Material | Silicone, Thermoplastic Elastomer |
| Product Type | Reusable Full Face Mask Respirator |
| Size | Small |
| Facepiece Type | Full |
| For Use With (Application) | For construction applications |
| For Use With (Equipment) | 200 series filters and 600 series cartridges |
| Harness Style | 4-point |
| Show More |