Learn More
Consort 6010 Multiparameter Portable Analyzer
Product Code. 12117810
Change view
Click to view available options
Product Type:
Analyzer with pH Kit
Portable Analyzer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
12117810 Portable Analyzer Each
15883182 Analyzer with pH Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 12117810 Supplier Consort Supplier No. C6010
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Suitable as bench-top and as portable device. Consort 6010 Multiparameter Portable Analyzer is a single-channel meter compatible with an array of electrodes that features an easy-to-use menu system and built-in help system to guide users through configuration and use.
Meter available separately or with pH kit (pH/ATC electrode SP20T, 2 x 50mL buffers (pH 4 and 7), 50mL electrolyte (3M KCl), and carrying case).
  • Portable and bench-top combined in one instrument
  • Parameters: pH, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen, temperature
  • Gold plated BNC connectors resist corrosion to ensure a long life, even in harsh environments
  • Pre-programmed pH buffers and EC standards, plus add up to 5 pH buffers and 3 EC standards
  • Custom calibration tables provide complete buffer/standard-temperature relation info (add via menu or from PC)
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings on mV, pH and DO
  • Range lock for conductivity measurements
  • Capacitive compensation eliminates electrode/cable capacitive component with low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • GPL report can be shown on the display or sent to the digital port
  • Pre-programmed standards
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be
Specifications
Outputs USB
Accuracy (Conductivity) 0.5% f.s. of range
Accuracy (DO) 1% ± 1 digit
Accuracy (mV) 0.1% ± 1 digit
Accuracy (pH) 0.1% ±1 digit
Backlight Option White backlight
Battery Type 4 x 1.2 V, AA, NiMH
Calibration GLP, reminder: 0 - 999 h
Calibration Points 1 (mV, DO, air pressure)
Calibration Points (Conductivity) 1 to 3
Calibration Points (pH) 1 to 5
Cell Constant 0.07 to 13 cm-1
Data Management Details All modes, manual or timed, interval: 1 - 9999 s
Data Points 12,000 + °C/date/time
Channels 1 measurement, 1 temperature
Display Type LCD, 128 x 64 pixel
Range (Conductivity) 0 to 2000 mS/cm
Range (mV) ±2000.0
Range (pH) -2.000 to 16.000
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0.0 to 70.0
Range (TDS) 0 to 100.0 g/L
Resolution (pH) 0.001
Temperature Range (Metric) -5°C to +105°C
Includes Meter Only
Inputs 1 BNC for measurement, 1 x 2 banana for temp
Languages English, Dutch, German, French
Model C6010
Power Supply 100/240 VAC, 50/60 Hz
Product Type Portable Analyzer
Range 600 to 1300 hPa (air pressure)
Range (RDO) 0 to 60.00 mg/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution (Conductivity) 0.001 μS/cm
Resolution (mV) 0.1
Resolution (RDO) 0.01 mg/L
Resolution (Resistivity) 1 Ω.cm
Resolution (TDS) 0.01 mg/L
Temperature Accuracy 0.1°C
Temperature Compensation −5° to +105° C (pH, conductivity), 0° to 50° C (DO)
Temperature (Metric) Operating 0°C to 40°C
Temperature Probe Calibration 1 point
Temperature Resolution 0.1° C
Temperature Sensor Pt1000
Warranty 36 Months
Dimensions (L x W x H) 12W x 25D x 5H cm
Weight (Metric) 600 g