missing translation for 'onlineSavingsMsg'
Learn More

(6E,10E)-7,11,15-Trimethyl-3-oxohexadeca-6,10,14-trienoic Acid, Ethyl Ester,(Mixture of Isomers), TRC

Product Code. 30457138
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457138 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457138 Supplier Toronto Research Chemicals Supplier No. TRCT796580500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6E,10E)-7,11,15-Trimethyl-3-oxohexadeca-6,10,14-trienoic Ac
CAS 141538-75-2
Molecular Formula C21 H34 O3
Quantity 500 mg
InChI Formula InChI=1S/C21H34O3/c1-6-24-21(23)16-20(22)15-9-14-19(5)13-8-12-18(4)11-7-10-17(2)3/h10,12,14H,6-9,11,13,15-16H2,1-5H3/b18-12+,19-14+
SMILES CCOC(=O)CC(=O)CC\C=C(/C)\CC\C=C(/C)\CCC=C(C)C
IUPAC Name ethyl (6E,10E)-7,11,15-trimethyl-3-oxohexadeca-6,10,14-trienoate
Molecular Weight (g/mol) 334.49
Formula Weight 334.2508
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.