missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 5192-04-1 |
| Melting Point | 96°C to 100°C |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass bottle |
| Molecular Formula | C8H8N2 |
| Quantity | 1 g |
| Synonym | 2-chloro-6-hydrazinopyridine, 2-chloro-6-hydrazinylpyridine, 6-chloro-2-pyridyl hydrazine, 6-chloro-2-pyridylhydrazine, 2 1h-pyridinone, 6-chloro-, hydrazone, pubchem12983, acmc-1awi0, 2-hydrazino-6-chloropyridine, 6-chloro-2-hydrazinopyridine, 6-chloro-2-hydrazonopyridine |
| Solubility Information | Solubility in water: insoluble |
| InChI Key | WTFWZOSMUGZKNZ-UHFFFAOYSA-N |
| SMILES | NC1=C2NC=CC2=CC=C1 |
| Show More |
Safety and Handling
Product Identifier
- 7-Aminoindole
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?