missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Azaindole, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 7-Azaindole |
| CAS | 271-63-6 |
| Molecular Formula | C7 H6 N2 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C7H6N2/c1-2-6-3-5-9-7(6)8-4-1/h1-5H,(H,8,9) |
| SMILES | c1cnc2[nH]ccc2c1 |
| IUPAC Name | 1H-pyrrolo[2,3-b]pyridine |
| Molecular Weight (g/mol) | 118.14 |
| Formula Weight | 118.0531 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction