missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxyindole, 98%, Thermo Scientific Chemicals

Catalog Number 11481120
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
20289-27-4
C15H13NO
223.28
MFCD00037974
DIGZMTAFOACVBW-UHFFFAOYSA-N
7-benzyloxyindole, 7-benzyloxy-1h-indole, 7-phenylmethoxy indole, 1h-indole, 7-phenylmethoxy, 7-phenylmethoxy-1h-indole, 7-benzyloxy-1h-indol, 7-benzyloxindole, 7-benzyloxy indole
260798
7-phenylmethoxy-1H-indole
C(OC1=C2NC=CC2=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
20289-27-4
C15H13NO
223.28
MFCD00037974
DIGZMTAFOACVBW-UHFFFAOYSA-N
7-benzyloxyindole, 7-benzyloxy-1h-indole, 7-phenylmethoxy indole, 1h-indole, 7-phenylmethoxy, 7-phenylmethoxy-1h-indole, 7-benzyloxy-1h-indol, 7-benzyloxindole, 7-benzyloxy indole
260798
7-phenylmethoxy-1H-indole
C(OC1=C2NC=CC2=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 74°C
Quantity 1 g
Formula Weight 223.28
Percent Purity 98%
Chemical Name or Material 7-Benzyloxyindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'