missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine, TRC

Product Code. 17992945
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992945 5 mg 5mg
17962175 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17992945 Supplier Toronto Research Chemicals Supplier No. TRCC3694455MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448950-89-8
Molecular Formula C17 H13 Cl N4
Molecular Weight (g/mol) 308.76
Synonym Alprazolam Imp. G (EP), 7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
IUPAC Name 7-chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
SMILES Cc1nnc2c(N)c(c3ccccc3)c4cc(Cl)ccc4n12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H13ClN4/c1-10-20-21-17-16(19)15(11-5-3-2-4-6-11)13-9-12(18)7-8-14(13)22(10)17/h2-9H,19H2,1H3
Formula Weight 308.0829
Percent Purity >95%
Chemical Name or Material 7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.