missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-4-oxo-4H-chromene-2-carboxylic Acid, TRC

Product Code. 30349736
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349736 10 mg
30366370 100 mg
30367499 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349736 Supplier Toronto Research Chemicals Supplier No. TRCC37562010MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114741-22-9
Molecular Formula C10H5ClO4
Molecular Weight (g/mol) 224.6
IUPAC Name 7-chloro-4-oxochromene-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)c2ccc(Cl)cc2O1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H5ClO4/c11-5-1-2-6-7(12)4-9(10(13)14)15-8(6)3-5/h1-4H,(H,13,14)
Formula Weight 223.9876
Chemical Name or Material 7-Chloro-4-oxo-4H-chromene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.