missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-5-phenyl-1,5-dihydro-3H-1,5-benzodiazepine-2,4-dione (Norclobazam), Mikromol™

Product Code. 30457929
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457929 4 x 25 mg -
17932986 100 mg 100mg
17942986 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457929 Supplier Mikromol Supplier No. MM0910.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-Chloro-5-phenyl-1,5-dihydro-3H-1,5-benzodiazepine-2,4-dione (Norclobazam), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethylclobazam
CAS 22316-55-8
Analyte or Component Names N-Desmethylclobazam
Shipping Condition Room Temperature
Molecular Formula C15 H11 Cl N2 O2
Certifications/Compliance ISO 9001
Synonym 8-chloro-1-phenyl-1H-1,5-benzodiazepine-2,4(3H,5H)-dione, 7-Chloro-5-phenyl-1,2,4,5-tetrahydro-3H-1,5-benzodiazepine-2,4-dione, Clofazin, Demethylclobazam, Desmethylclobazam, N-Demethylclobazam, N-Desmethylclobazam, Norclobazam, Clobazam Imp. A (EP), Clobazam Impurity A
InChI Formula InChI=1S/C15H11ClN2O2/c16-10-6-7-12-13(8-10)18(11-4-2-1-3-5-11)15(20)9-14(19)17-12/h1-8H,9H2,(H,17,19)
Solution Type Neat
SMILES Clc1ccc2NC(=O)CC(=O)N(c3ccccc3)c2c1
IUPAC Name 7-chloro-5-phenyl-1H-1,5-benzodiazepine-2,4-dione
Molecular Weight (g/mol) 286.71
Quantity 4 x 25 mg
Formula Weight 286.0509 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.