missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroindole, 98%, Thermo Scientific™

Product Code. 10752054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10752054 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10752054 Supplier Thermo Scientific Acros Supplier No. 391340010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53924-05-3
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 86°C to 88°C (5.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H6ClN
MDL Number MFCD01321301
Quantity 1g
Synonym 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq
InChI Key WMYQAKANKREQLM-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C1)Cl)NC=C2
IUPAC Name 7-chloro-1H-indole
Molecular Weight (g/mol) 151.6
PubChem CID 104644
Formula Weight 151.6
Percent Purity 98%
Chemical Name or Material 7-Chloroindole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.