missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, 98+%, Thermo Scientific™

NMDA receptor antagonist at the glycine site

87.79€ - 322.01€

Chemical Identifiers

CAS 18000-24-3
Molecular Formula C10H6ClNO3
Molecular Weight (g/mol) 223.61
MDL Number MFCD00069227
InChI Key UAWVRVFHMOSAPU-UHFFFAOYSA-N
Synonym 7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
PubChem CID 1884
ChEBI CHEBI:63965
IUPAC Name 7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403969
SDS View Documents
Thermo Scientific Alfa Aesar
J61359.MA
10mg
ONLINE EXCLUSIVE PRICE
89.60€
87.79€ / 10mg
Save 1.81€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15413969
SDS View Documents
Thermo Scientific Alfa Aesar
J61359.MF
50mg
ONLINE EXCLUSIVE PRICE
328.00€
322.01€ / 50mg
Save 5.99€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18000-24-3
223.61
UAWVRVFHMOSAPU-UHFFFAOYSA-N
1884
7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
C10H6ClNO3
MFCD00069227
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
CHEBI:63965
OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1

Specifications

18000-24-3
MFCD00069227
Soluble in DMSO or dilute sodium hydroxide
OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1
223.61
CHEBI:63965
≥99%
C10H6ClNO3
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
UAWVRVFHMOSAPU-UHFFFAOYSA-N
7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
1884
223.62
7-Chlorokynurenic acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 241-913-6

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.