missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)coumarin-3-carbonyl Azide, TRC

Product Code. 30180415
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180415 50 mg 50mg
30180412 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180415 Supplier Toronto Research Chemicals Supplier No. TRCD47372850MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157673-16-0
Molecular Formula C19 H19 Cl N6 O
Molecular Weight (g/mol) 382.847
Synonym 7-(Diethylamino)-2-oxo-2H-1-benzopyran-3-carbonyl Azide
IUPAC Name 5-[(8-chloroisoquinolin-3-yl)amino]-3-[(2R)-1-(dimethylamino)propan-2-yl]oxypyrazine-2-carbonitrile
SMILES C[C@H](CN(C)C)Oc1nc(Nc2cc3cccc(Cl)c3cn2)cnc1C#N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H19ClN6O/c1-12(11-26(2)3)27-19-16(8-21)22-10-18(25-19)24-17-7-13-5-4-6-15(20)14(13)9-23-17/h4-7,9-10,12H,11H2,1-3H3,(H,23,24,25)/t12-/m1/s1
Formula Weight 382.131
Chemical Name or Material 7-(Diethylamino)coumarin-3-carbonyl Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.