missing translation for 'onlineSavingsMsg'
Learn More

7-Ethylindole, 98+%

Product Code. 11313716
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313716 10 g 10g
11323716 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313716 Supplier Thermo Scientific Alfa Aesar Supplier No. L14875.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22867-74-9 | C10H11N | 145.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22867-74-9
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
MDL Number MFCD00143514
InChI Key PIIZLMYXLGYWTN-UHFFFAOYSA-N
Synonym 7-ethylindole, 1h-indole, 7-ethyl, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
PubChem CID 2724711
IUPAC Name 7-ethyl-1H-indole
SMILES CCC1=C2NC=CC2=CC=C1

Specifications

Density 1.058
Boiling Point 230°C
Refractive Index 1.603
Quantity 10 g
Beilstein 1238356
Formula Weight 145.21
Percent Purity ≥98%
Chemical Name or Material 7-Ethylindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.