missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Fluphenazine, TRC

Product Code. 30009702
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009702 1 mg 1mg
30004571 5 mg 5mg
30003478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009702 Supplier Toronto Research Chemicals Supplier No. TRCH9424351MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33098-48-5
Molecular Formula C22 H26 F3 N3 O2 S
Molecular Weight (g/mol) 453.52
Synonym 10-[3-[4-(2-Hydroxyethyl)-1-piperazinyl]propyl]-8-(trifluoromethyl)-10H-phenothiazin-3-ol, 4-[3-[7-Hydroxy-2-(trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazineethanol;
IUPAC Name 10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]-8-(trifluoromethyl)phenothiazin-3-ol
SMILES OCCN1CCN(CCCN2c3ccc(O)cc3Sc4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C22H26F3N3O2S/c23-22(24,25)16-2-5-20-19(14-16)28(18-4-3-17(30)15-21(18)31-20)7-1-6-26-8-10-27(11-9-26)12-13-29/h2-5,14-15,29-30H,1,6-13H2
Formula Weight 453.1698
Percent Purity >95%
Chemical Name or Material 7-hydroxy Fluphenazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.