missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Methyl-4-indanol, 97%

Product Code. 11449876
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449876 10 g 10g
11439876 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449876 Supplier Alfa Aesar™ Supplier No. B20343.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
7-Methyl-4-indanol is used as an intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16400-13-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00173675
InChI Key XHMLXYGITDAGDN-UHFFFAOYSA-N
Synonym 7-methylindan-4-ol, 4-hydroxy-7-methylindane, 1h-inden-4-ol,2,3-dihydro-7-methyl, 7-methyl-4-indanol, acmc-20amgr, 7-methyl-indan-4-ol
PubChem CID 85398
IUPAC Name 7-methyl-2,3-dihydro-1H-inden-4-ol
SMILES CC1=CC=C(O)C2=C1CCC2

Specifications

Melting Point 80°C to 82°C
Quantity 10 g
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 7-Methyl-4-indanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.