missing translation for 'onlineSavingsMsg'
Learn More

7-Methylbenzo[d]isoxazol-3(2H)-one, 98%, BLD Pharm™

Product Code. 30987344
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30987344 5 g
30971497 1 g
30980826 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30987344 Supplier BLD Pharm Supplier No. BD1108615g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 36238-83-2
Molecular Formula C8H7NO2
MDL Number MFCD00104669
Quantity 5 g
SMILES CC1=C2ONC(=O)C2=CC=C1
Molecular Weight (g/mol) 149.15
Percent Purity 98%
Chemical Name or Material 7-Methylbenzo[d]isoxazol-3(2H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.