Learn More
Uvex™ 7310 silv-Air Pro FFP3 Breathing Mask
Product Code. 10892041 Shop All Uvex Products
Change view
Click to view available options
:
FFP3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10892041 FFP3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10892041 Supplier Uvex™ Supplier No. 8707310
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Mask provides protection against pollutants up to 30 times the limit value, with an exhalation valve to reduce breathing resistance and heat under the mask.
  • Protection against pollutants up to 30 times the limit value
  • 4-point headband attachment for a comfortable, individual and secure fit of the mask
  • Integrated , flexibly adjustable nose clip
  • Exhalation valve reduces breathing resistance and heat/moisture under the mask
  • Very soft, circumferential sealing lip for optimal wearing comfort and a reliable sealing fit
  • Mask meets the requirements for reusability (R), and against high dust exposure (D marking, dolomite dust test)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Color Gray
With Exhalation Valve Yes
Filter Efficiency FFP3
Model 7310 silv-Air Pro