Learn More
Knick Elektronische™ 765 Waterproof Laboratory pH Meter
Product Code. 10440833
Change view
Click to view available options
Type:
pH Meter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10440833 pH Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10440833 Supplier Knick Elektronische™ Supplier No. 765
  • ONLINE EXCLUSIVE PRICE
    3050.25€
    2332.82€ / Pack of 1
    Save 717.43€
    23% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Equipped with an exemplary package of safety functions. 765 Waterproof Laboratory pH Meter is reliable and makes taking pH measurements easier than ever.
  • Fullcheck automatic device test
  • Records for QM documentation according to ISO 9000 and GLP
  • Trueline calibrated analog recorder output, galvanically isolated
  • Sensor monitoring with Sensoface icon alerts
  • Automatic calibration with patented Calimatic
  • EMC according to NAMUR
  • Sensor statistics
  • RS-232 interface for computer and printer
  • Displaymatic
  • Two user-defined measured values displayed simultaneously
  • Dead-stop operation
  • Self-contained clock
  • Leak-proof membrane keyboard
  • Robust housing
  • IP 54 protection
  • 3-year warranty
Specifications
Includes Real-Time Clock with Date, Automatic Data Storage, Power Cord
For Use With (Application) Reliable, simple pH measurements
Calibration Automatic
Keypad Leak-proof membrane keyboard
Model 765 Laboratory pH Meter
Type pH Meter