Learn More
8-Bromo-6-cyanoimidazo[1,2-a]pyridine, 95%, Thermo Scientific™
Product Code. 15457457
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457457 250 mg 250mg
15447457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15457457 Supplier Thermo Scientific Alfa Aesar Supplier No. H35030.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1221791-93-0
Molecular Formula C8H4BrN3
Molecular Weight (g/mol) 222.045
MDL Number MFCD14584821
InChI Key DWOXNHVFPVGULV-UHFFFAOYSA-N
Synonym 8-bromoimidazo 1,2-a pyridine-6-carbonitrile, 8-bromo-6-cyanoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-carbonitrile,8-bromo, 8-bromo-4-hydroimidazo 1,2-a pyridine-6-carbonitrile
PubChem CID 50853181
IUPAC Name 8-bromoimidazo[1,2-a]pyridine-6-carbonitrile
SMILES C1=CN2C=C(C=C(C2=N1)Br)C#N
Specifications
Melting Point 184°C to 186°C
Quantity 250 mg
Formula Weight 222.05
Percent Purity 95%
Chemical Name or Material 8-Bromo-6-cyanoimidazo[1,2-a]pyridine