missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyquinoline, 96%, Thermo Scientific Chemicals

Catalog Number 15410738
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
938-33-0
C10H9NO
159.19
MFCD00957068
ZLKGGEBOALGXJZ-UHFFFAOYSA-N
quinoline, 8-methoxy, methyl 8-quinolyl ether, ccris 7821, unii-4y56u1unuj, 4y56u1unuj, 8-methoxyquinoline, 8-methoxy quinoline, 8-methoxy-quinoline, acmc-209rns, ksc268q2n
70310
8-methoxyquinoline
COC1=C2N=CC=CC2=CC=C1
This item is not returnable. View return policy
938-33-0
C10H9NO
159.19
MFCD00957068
ZLKGGEBOALGXJZ-UHFFFAOYSA-N
quinoline, 8-methoxy, methyl 8-quinolyl ether, ccris 7821, unii-4y56u1unuj, 4y56u1unuj, 8-methoxyquinoline, 8-methoxy quinoline, 8-methoxy-quinoline, acmc-209rns, ksc268q2n
70310
8-methoxyquinoline
COC1=C2N=CC=CC2=CC=C1

8-Methoxyquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methoxyquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 41°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 159.19
Percent Purity 96%
Chemical Name or Material 8-Methoxyquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'